Metzenbaum Scissors
Product Details & Documents
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue.
The tips are semi-blunt, and they are available in curved or straight styles.
Specifications
- Description:Metzenbaum scissors, straight tips
- Length:14.5 cm (5.75")
- Material:Stainless steel