Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
NEXTFLEX™ UDI-UMI Barcodes

NEXTFLEX™ UDI-UMI Barcodes

NEXTFLEX™ UDI-UMI barcodes (1 - 24), 24 reactions
Catalog # 77660-622
Supplier #:  NOVA-734101
NEXTFLEX™ UDI-UMI Barcodes
Catalog # 77660-622
Supplier:  
CAS Number:  
NEXTFLEX™ UDI-UMI Barcodes

NEXTFLEX™ UDI-UMI barcodes (1 - 24), 24 reactions

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The NEXTFLEX™ UDI-UMI Barcodes consist of 96 unique dual-index (UDI) barcodes, each with a 10-base pair UDI and a 9-base pair unique molecular identifier (UMI). These adapters are designed for multiplexing both DNA and RNA NGS libraries on all Illumina® and Element® sequencers and are compatible with any TruSeq® style library prep kit.

  • Set of 96 full length UDI (10 bp) - UMI (9 bp) barcodes for multiplexing NGS libraries for Illumina® and Element sequencers
  • Compatible with Revvity library prep and 'T-A' overhang (Illumina’s TruSeq style barcodes) ligation-based workflows
  • Compatible with PCR-free workflows
  • Every lot is functionally verified and tested for index purity by sequencing

Unique dual indexes (UDIs) and Unique molecular identifiers (UMIs) are nucleotide sequences or 'barcodes' that are incorporated during library preparation for Next-generation sequencing (NGS) and provide several advantages.

Unique dual indexes (UDIs): Revvity's 10 bp UDI are incorporated on both ends of an NGS library molecule, enabling researchers to sequence multiple samples in parallel, which is often referred as multiplexing. The presence of two indices minimizes the effects of index hopping, a phenomenon where a read is assigned to the wrong index as consequence of errors that appear during sequencing process. UMIs are also referred as 'Molecular Barcodes' or 'Random Barcodes'. The presence of UMI allows differentiation of PCR duplicates (which contain same UMI sequence) from true copy number (each copy contains a different UMI sequence). They are an excellent tool to assess library complexity. UMI sequence information in conjunction with alignment coordinates enables grouping of sequencing data into read families representing individual sample DNA or RNA fragments. UMIs are used mainly in the context of quantitative sequencing and rare variant analysis.

Quantitative sequencing: When starting from low input samples, stochastic effects in the first rounds of the PCR and the biases of the PCR polymerases against GC content can potentially cause erroneous quantitation data. Removal of PCR duplicates using alignment coordinate information is known to be inefficient for low input situations. It is also known to be inefficient for deep sequencing of RNA-seq or ChIP-Seq libraries (>80 million reads per sample). In the latter case alignment coordinate-based de-duplication will remove large numbers of biological duplicate reads, especially for the most abundant transcripts. As indicated above, UMIs alleviate the PCR duplicate problem by adding unique molecular tags to the sequencing library molecules before amplification.

Rare variant analysis: NGS sequencing provides data with low error rates (~0.1 to 0.5%) for most applications. These low error rates interfere with the confident identification of low abundance variants. UMI-less data can’t distinguish between these and sequencing errors. By incorporating individual barcodes on each original DNA fragment, variant alleles present in the original sample (true variants) can be distinguished confidently from errors introduced during library preparation, target enrichment, or sequencing. Any identified errors can be removed by bioinformatics methods before final data analysis. Applications include sequencing of heterogeneous tumor samples, cfDNA sequencing including ctDNA sequencing and deep exome sequencing.

The incorporation of full-length UDI and UMI sequences by ligation instead of PCR reduces the possibility of introducing errors and allows taking advantage of these features in PCR-free workflows. For enhanced variant calling, gene expression analysis, sequencing of heterogeneous tumor samples, ctDNA sequencing, deep exome sequencing, single-cell RNA-seq, and haplotyping via linked reads, we suggest utilizing the NEXTFLEX™ UMI-UDI barcodes. In summary, UMIs provide error correction, enhance variant calling, and improve overall data in genomics research.

Certifications: Manufactured in ISO 9001 facility.

Caution: For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    NEXTFLEX™ UDI-UMI barcodes (1 - 24)
  • No. of reactions:
    24 reactions

Specifications