Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

GBS Detect/Strep Reagents

GBS Detect™, for non-hemolytic group B Strep, 15×100 mm plate, 100 plates/box
Catalog # 77623-656
Supplier:   Hardy Diagnostics
 :  A300BX
GBS Detect/Strep Reagents
Catalog # 77623-656
Supplier:  Hardy Diagnostics
CAS Number:  
GBS Detect/Strep Reagents
GBS Detect/Strep Reagents

GBS Detect™, for non-hemolytic group B Strep, 15×100 mm plate, 100 plates/box

CAS Number:  
Price
$376.00
Log in for availability
Pack of 100
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

GBS Detect is used for the isolation and differentiation by enhanced hemolysis of gamma hemolytic Group B Streptococcus.

  • Grouping reagents for the rapid latex agglutination test
  • Lancefield grouping includes A,B,C,D,F,G latex, controls, extraction reagents, cards, and mixing sticks
  • StrepQuick™ to aid in the identification of gram positive cocci

GBS Detect™, for non-hemolytic Group B Strep. Granada Medium for hemolytic Group B Strep. Grouping Reagents A, B, C, and G, for the rapid latex agglutination test. PYR Test Kit with chromogenic developer on paper disks. Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), especially for throat cultures. Selective Strep Agar (COBA Medium), for the selective isolation of all species of Strep. ci. StrepQuick™ performs a PYR, LAP, and Esculin test to aid in the identification of gram positive coc.

Specifications

Specifications

  • Description:
    GBS Detect™, for non-hemolytic group B Strep
  • Size:
    100 plates/box
  • Format:
    15×100 mm plate

Specifications