Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER TRIS SALINE-TWEEN PH7.4 4X1L
Catalog # MSPP-IBB181R4X
Supplier:
BOSTON BIOPRODUCTS MS
Expand image
BUFFER TRIS SALINE-TWEEN PH7.4 4X1L
Catalog # MSPP-IBB181R4X
Supplier:
BOSTON BIOPRODUCTS MS
Supplier Number:
IBB-181R-4X1L