Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

VWR® pH Electrodes

pH composite electrode (Slender), Glass, S7 [S7-BNC(Q9) cable], 3 M KCl, 0...60 °C, 0 - 14
Catalog # 77618-878
Supplier:   VWR International
VWR® pH Electrodes
Catalog # 77618-878
Supplier:  VWR International
VWR® pH Electrodes
VWR® pH Electrodes

pH composite electrode (Slender), Glass, S7 [S7-BNC(Q9) cable], 3 M KCl, 0...60 °C, 0 - 14

Price
$463.95
Log in for availability
Each
Product Details & Documents

The VWR® collection pH electrodes provide a wide-array of sensor options for any application requirement.

  • Specialized sensor configurations for various applications including wastewater, soil, food and beverage, and micro-container measurements

  • BNC connector (BNC(Q9) or S7)
  • Variety of body materials including glass, lead-free glass, polycarbonate, and epoxy
  • Gel or liquid electrolyte fill solution
  • Includes unique serial number
  • Easily connect to VWR(R) pH meters

For additional specifications and application specific recommendations, please reference 'VWR® pH Electrode Selection Guide' located in the Documents tab below.

Documents

Specifications
  • Temperature range:
    0...60 °C
  • Type:
    pH composite electrode (Slender)
  • Material:
    Glass
  • pH range:
    0 - 14
  • Fill solution:
    3 M KCl
  • Connector:
    S7 [S7-BNC(Q9) cable]
Accessories

Product Information

  • Catalog #
    Material
    For use with
    Availability
    Price
  • VWR® ATC probe
    VWR® ATC probe
    77618-918
    316 stainless steel
    VWR® 500-, 550-, 700-, 750-, 900, 950-series meters
    Log in for availability
    Each - $129.12

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.