Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER 2357 (8 ENVELOPES-4 LITERS)
Catalog # MSPP-98005
Supplier:
YSI INC. MS
Expand image
BUFFER 2357 (8 ENVELOPES-4 LITERS)
Catalog # MSPP-98005
Supplier:
YSI INC. MS
Supplier Number:
98005