Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TAG PRE-WIRED 3-3/4 X 1-7/8IN PK1000
Catalog # MSPP-S-1151PW
Supplier:
ULINE MS
Expand image
TAG PRE-WIRED 3-3/4 X 1-7/8IN PK1000
Catalog # MSPP-S-1151PW
Supplier:
ULINE MS
Supplier Number:
S-1151PW