Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TAG SHIPPNG 1 PLAIN 2-3/4X1-3/8IN PK1000
Catalog # MSPP-S1474PLAN
Supplier:
ULINE MS
Expand image
TAG SHIPPNG 1 PLAIN 2-3/4X1-3/8IN PK1000
Catalog # MSPP-S1474PLAN
Supplier:
ULINE MS
Supplier Number:
S-1474PLAIN