BUFFER RIPA W/TRITON-X 250ML
Catalog # MSPP-BP-116TX
Supplier: BOSTON BIOPRODUCTS MS
BUFFER RIPA W/TRITON-X 250ML
Catalog # MSPP-BP-116TX
Supplier: BOSTON BIOPRODUCTS MS
Supplier Number: BP-116TX-250ML
Frequently Bought Together
Vi-CELL BLU Reagent Kits, Beckman Coulter
Supplier: Beckman Coulter
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Disposable PD-10 Desalting Columns, Cytiva
Supplier: Cytiva
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
VWR® Disposable Serological Pipettes
Supplier: VWR International
Biox Cell Scrapers and Lifters
Supplier: TWD Scientific
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: AB SCIEX LLC MS
BUFFER TBS BLOCKING READY-TO-USE 500ML
Supplier: Li-Cor
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza