Polystyrene Work Racks/Containers
Work racks/containers, 5×4 format, White
Catalog # 77589-898
Supplier: SARSTEDT INC
Supplier #: 95.64.259
Jump to:
Product Details & Documents
High-quality, safe, and robust containers are essential for specimen processing and storage. SARSTEDT offers the right product for every laboratory and analytical requirement.
- Work rack is cost-effective and saves space
- Lid ensures secure sample storage
- Various designs for tubes up to max. Ø 16 mm
SARSTEDT's practical and versatile range of racks and storage boxes provides a solution for every application.
Specifications
- Color:White
- Description:Work racks/containers, 5×4 format