$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
RACK F/SCREW CAP HIGH COVER 48-4 CS10
MSPP-MP51301
:
NBS SCIENTIFIC, LLC MS
RACK F/SCREW CAP HIGH COVER 48-4 CS10
MSPP-MP51301
:
NBS SCIENTIFIC, LLC MS
:
MP51301