Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CAP SCREW INTERN THREAD TUBE GREY CS10
Catalog # MSPP-MP53191
Supplier:
NBS SCIENTIFIC, LLC MS
Expand image
CAP SCREW INTERN THREAD TUBE GREY CS10
Catalog # MSPP-MP53191
Supplier:
NBS SCIENTIFIC, LLC MS
Supplier Number:
MP53191