Inoculation Loops
Inoculation loop, 1 µl, White, Blister pack
Catalog # 77589-576
Supplier: SARSTEDT INC
Supplier #: 86.1567.050
Jump to:
Product Details & Documents
One aspect of microbiology is researching and identifying pathogens, as well as looking for suitable treatments. SARSTEDT provides researchers with the consumables they need to do their work.
- Material: PS
- Facilitate swabbing on a Petri dish with a culture medium
- No damage to the culture medium thanks to the smooth and even surface
- Save time when processing larger test series
- Safe handling - no pathogenic aerosols or cross contamination
- Sterilized by gamma irradiation
Flexible SARSTEDT inoculation loops allow swabs to be taken easily or inoculation in liquid media. The inoculation needle can be used to inoculate or remove individual colonies. We recommend use of the inoculation spreader to evenly apply larger volumes to culture media.
Specifications
- Volume:1 µl
- Color:White
- Description:Inoculation loop
- Packaging:Blister pack