Reaction Tubes without Cap
Reaction tubes without cap, Transparent, 2 ml
Catalog # 77588-934
Supplier: SARSTEDT INC
Supplier #: 72.708
Jump to:
Product Details & Documents
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Polypropylene tubes supplied without closures
- Particularly suitable for purification steps and aliquoting processes
- Good visibility of the sample
- Surface offers sufficient space for labelling
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base; with or without skirted base; with push cap or screw cap; with or without print or labels.
Specifications
- Volume:2 ml
- Color:Transparent
- Description:Reaction tubes without cap
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



