Low Binding Reaction Tubes
Low DNA-binding surface, Transparent, 2 ml, Transparent, PP, Non sterile (PCR performance tested)
Catalog # 77588-854
Supplier: SARSTEDT INC
Supplier #: 72.695.700
Jump to:
Product Details & Documents
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- SafeSeal polypropylene caps attached
- Reduced binding properties for higher recovery rates
- PCR Performance Tested certified
- Secure seal even under thermal and mechanical stress
- High centrifugation up to at least 20,000 ×g
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base, with or without skirted base, with push cap or screw cap, with or without print or labels.
Specifications
- Cap type:Transparent, PP
- Sterility:Non sterile (PCR performance tested)
- Volume:2 ml
- Color:Transparent
- Description:Low DNA-binding surface