Containers with Assembled Screw Cap
120 ml, Neutral, Transparent, Non sterile
Catalog # 77589-070
Supplier: SARSTEDT INC
Supplier #: 75.9922.421
Jump to:
Product Details & Documents
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Flat base shape
- Polypropylene
- Volume can be clearly identified via graduated scale
- Tamper-evident seal on the collection tube
SARSTEDT collection cups and containers are available in a range of sizes and volumes.
Specifications
- Sterility:Non sterile
- Capacity:120 ml
- Color:Transparent
- Cap color:Neutral