Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Reaction Tubes

Reaction Tubes

Reaction tube, Blue, 1.5 ml
Catalog # 77588-740
Supplier:   SARSTEDT INC
Supplier #:  72.690.375
Reaction Tubes
Catalog # 77588-740
Supplier:  SARSTEDT INC
Reaction Tubes
Reaction Tubes

Reaction tube, Blue, 1.5 ml

Price
$32.81
Log in for availability
Pack of 500 Case of 5000
Product Details & Documents

High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • Polypropylene retaining loop caps attached
  • Polypropylene tube material
  • Easy to open, secure seal when closed
  • Larger packaging units for economical storage and stocking
  • High centrifugation up to 20000×g

This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base; with or without skirted base; with push cap or screw cap; with or without print or labels.

Assorted color options contain a mix of red, green, blue, purple, yellow and neutral.

Specifications
  • Volume:
    1.5 ml
  • Color:
    Blue
  • Description:
    Reaction tube
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.