Reaction Tubes
Reaction tube, Brown, 1.5 ml
Catalog # 77588-736
Supplier: SARSTEDT INC
Supplier #: 72.690.304
Jump to:
Product Details & Documents
High-quality, safe, and robust containers are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Polypropylene retaining loop caps attached
- Polypropylene tube material
- Easy to open, secure seal when closed
- Larger packaging units for economical storage and stocking
- High centrifugation up to 20000×g
This broad product portfolio in the field of test and micro tubes includes options with round, conical or flat base; with or without skirted base; with push cap or screw cap; with or without print or labels.
Assorted color options contain a mix of red, green, blue, purple, yellow and neutral.
Specifications
- Volume:1.5 ml
- Color:Brown
- Description:Reaction tube



