Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cuvettes

Cuvettes

Cuvettes, Rack pack, PS
Catalog # 77588-360
Supplier:   SARSTEDT INC
Supplier #:  67.741
Cuvettes
Catalog # 77588-360
Supplier:  SARSTEDT INC
Cuvettes
Cuvettes

Cuvettes, Rack pack, PS

Price
$18.53
Log in for availability
Pack of 100 Case of 2000
Product Details & Documents

SARSTEDT has been known as a manufacturer of high-quality disposable cuvettes.

  • Height: 45 mm
  • Optical sides: 2
  • Path length: 10 mm
  • Disposable cuvette made of high-quality, crystal-clear polystyrene or acrylic (PMMA)
  • Ideal for photometric analyses of liquids in the wavelength range ≥300 nm (PMMA)/≥330 nm (PS)
  • An arrow integrated in the cuvette allows exact positioning in the photometer
  • Nest sorting ensures consistent transmission values within a polystyrene box

SARSTEDT cuvettes are made from polystyrene (PS), acrylic (PMMA) and a special plastic for analysis in the UV range, which are ideal for photometric analysis of liquid solutions. The cuvettes are also available in 2× optical semi-micro and a 4× optical format for 90° angle fluorescence measurements.

Specifications
  • Description:
    Cuvettes
  • Material:
    PS
  • Packaging:
    Rack pack
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.