Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Screw Cap Tubes with Skirted Round Base, PP

Screw Cap Tubes with Skirted Round Base, PP

PP screw cap tubes with skirted round base, with enclosed cap, PP, 16 mm×79 mm, 10 ml, Brown, Graduated, 500 pieces/bag, Non sterile
Catalog # 77587-952
Supplier:   SARSTEDT INC
Supplier #:  62.551.022
Screw Cap Tubes with Skirted Round Base, PP
Catalog # 77587-952
Supplier:  SARSTEDT INC
Screw Cap Tubes with Skirted Round Base, PP
Screw Cap Tubes with Skirted Round Base, PP

PP screw cap tubes with skirted round base, with enclosed cap, PP, 16 mm×79 mm, 10 ml, Brown, Graduated, 500 pieces/bag, Non sterile

Price
$282.98
Log in for availability
Case of 1,000
Product Details & Documents

High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.

  • Base shape: Skirted round base
  • Tube with skirted base ensures that it stands securely
  • Polypropylene (PP) tubes for thermal, mechanical and chemical applications
  • Especially secure cap
  • Corresponds to the requirement: IATA

This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.

Specifications
  • Ø×L:
    16 mm×79 mm
  • Sterility:
    Non sterile
  • Volume:
    10 ml
  • Description:
    PP screw cap tubes with skirted round base, with enclosed cap
  • Material:
    PP
  • Packaging:
    500 pieces/bag
  • Cap color:
    Brown
  • Graduations:
    Graduated