Screw Cap Tubes with Skirted Conical Base, PS
Screw cap tubes with skirted conical base, cap enclosed, Transparent, 10 ml, 16 mm, 97 mm, Yes, Natural, HDPE, Screw cap, 4000×g, Bulk, Non sterile
77587-832
: 60.9921.821
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, this range offers the right product for every laboratory and analytical requirement.
- Material: PS
- Graduations: No
- Base shape: Skirted conical base
- Corresponds to the requirement: ADR
- Tube with skirted base ensures that it stands securely
- Polystyrene (PS) tubes for maximum transparency
- Especially secure cap
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.
- Cap included:Yes
- Cap material:HDPE
- Cap type:Screw cap
- Ø:16 mm
- Sterility:Non sterile
- Volume:10 ml
- Color:Transparent
- Description:Screw cap tubes with skirted conical base, cap enclosed
- Length:97 mm
- Packaging:Bulk
- Max. rcf:4000×g
- Cap color:Natural



