Screw Cap Tubes with Skirted Round Base, PP
PP screw cap tubes with skirted round base, without cap, PP, 16.5 mm×101 mm, 13 ml, —, Non-Graduated, 500 pieces/bag, Non sterile
Catalog # 77587-730
Supplier: SARSTEDT INC
Supplier #: 60.541.009
Jump to:
Product Details & Documents
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- Base shape: Skirted round base
- Tube with skirted base ensures that it stands securely
- Polypropylene (PP) tubes for thermal, mechanical and chemical applications
- Especially secure cap
- Corresponds to the requirement: IATA
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.
Specifications
- Ø×L:16.5 mm×101 mm
- Sterility:Non sterile
- Volume:13 ml
- Description:PP screw cap tubes with skirted round base, without cap
- Material:PP
- Packaging:500 pieces/bag
- Cap color:—
- Graduations:Non-Graduated
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



