Screw Caps
Screw Cap, 13 mm, Dark blue
Catalog # 77588-070
Supplier: SARSTEDT INC
: 65.163.308
Jump to:
Product Details & Documents
High-quality, safe, and robust containers and caps are essential for mixing, processing, and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, SARSTEDT offers the right product for every laboratory and analytical requirement.
- HDPE and PP materials
- Especially secure cap for fluctuations in temperature
- Ribbed surface for maximum grip
Screw caps offer excellent sealing properties, and are available in a wide variety of colors to facilitate organization and coding.
Specifications
- Description:Screw Cap
- For tube diameter:13 mm
- Cap color:Dark blue