Detectable Forceps, Sterile
Product Details & Documents
These samplers are not only colored blue but also have a special additive in the material that makes them visible to metal detectors or X rays. They can, therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic. These disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example, fine tissue, small items or solid samples.
- PS detectable, blue
- Individually packed and sterilized
- For single use
- Cleanroom manufactured (class seven)
- Overall length 130 mm
The fluted handles ensure optimum and secure handling as well as precise work. These light plastic single-use forceps for allow for fatigue-free work. The cleanroom manufactured lab assistants meet the highest standards preventing cross-contamination or impurities in the sample being examined.
These lab forceps are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses.
Pointed/angled shape: These disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends.
Pointed shape: These anatomical forceps have a straight tip, fluted on the inside making it possible to grip sensitive materials reliably.
Broad tip shape: These disposable forceps have ends with a broad tip. The broad tip is smooth and blunt preventing the material from being damaged when gripped.
Certifications: Complies with EU food requirements and FDA regulations.
Documents
Specifications
- Color:Blue
- Description:Forecep detectable with straight type
- Length:130 mm
- Packed:Individually
- Sterile:Sterile