FreezerPro® Sample Management Software
FreezerPro® Sample management software solution, 4 h customized training session
Catalog # 77581-278
Supplier: AZENTA US, INC
: 001-TRD-100
Jump to:
Product Details & Documents
FreezerPro® is a web-based sample management inventory system designed to support organizations of all sizes with simple and quick installation, from lower throughput labs to central biorepositories handling millions of records.
- Delivers secure management of samples and sample information
- Register samples and generate automated or manual pick lists through a unified interface
- Track samples in and out of freezers
- Reporting and data export/import (multiple formats)
- Live and smart search: Quickly search for your samples by any property
- Full audit trails
FreezerPro® is an off-the-shelf sample inventory management software that provides clear visibility to all samples. The FreezerPro® user interface (UI) is standardized, simple, and intuitive, allowing for deployment within hours and rapid user adoption. FreezerPro® can be integrated with the cryogenic storage and Ziath tube and rack barcode reading systems from Azenta. Cloud-based and on-premises installations are available.
Specifications
- Description:FreezerPro® Sample management software solution, 4 h customized training session