Orders LinkContactUsLinkComponent
FreezerPro® Sample Management Software
FreezerPro® Sample Management Software
FreezerPro® Sample management software solution, cloud hosted, concurrent license fee per year
Catalog # 77581-274
Supplier:   AZENTA US, INC
Supplier #:  001-FPD-222
FreezerPro® Sample Management Software
Catalog # 77581-274
Supplier:  AZENTA US, INC
FreezerPro® Sample Management Software
FreezerPro® Sample Management Software
FreezerPro® Sample management software solution, cloud hosted, concurrent license fee per year
Price
$3,642.98
Log in for availability
Each
Product Details & Documents
FreezerPro® is a web-based sample management inventory system designed to support organizations of all sizes with simple and quick installation, from lower throughput labs to central biorepositories handling millions of records.
  • Delivers secure management of samples and sample information
  • Register samples and generate automated or manual pick lists through a unified interface
  • Track samples in and out of freezers
  • Reporting and data export/import (multiple formats)
  • Live and smart search: Quickly search for your samples by any property
  • Full audit trails
FreezerPro® is an off-the-shelf sample inventory management software that provides clear visibility to all samples. The FreezerPro® user interface (UI) is standardized, simple, and intuitive, allowing for deployment within hours and rapid user adoption. FreezerPro® can be integrated with the cryogenic storage and Ziath tube and rack barcode reading systems from Azenta. Cloud-based and on-premises installations are available.
Specifications
  • Description:
    FreezerPro® Sample management software solution, cloud hosted, concurrent license fee per year