Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
RecombiMAb Mouse IgG2a Isotype control Unknown Specificity

RecombiMAb Mouse IgG2a Isotype control Unknown Specificity

100 mg
Catalog # 77580-350
Supplier:   Bio X Cell
Supplier #:  CP160-100MG
RecombiMAb Mouse IgG2a Isotype control Unknown Specificity
Catalog # 77580-350
Supplier:  Bio X Cell
RecombiMAb Mouse IgG2a Isotype control Unknown Specificity

100 mg

Price
$6,488.87
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Type: Control
Antigen: IgG1 ?
Clonality: Recombinant
Clone: MOPC-21-CP160
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:

Specifications
  • Size:
    100 mg
  • Cat. No.:
    77580-350
  • Clonality:
    Recombinant
  • Flow Cytometry:
    Yes
  • Western Blot:
    Yes
  • Antibody Type:
    Control
  • Antigen Symbol:
    IgG1 ?
  • Tested Applications:
    in vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody
  • Clone:
    MOPC-21-CP160