Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Re-Usable Cleaning Coupons

Re-Usable Cleaning Coupons

Stainless steel 316L - satin, 50×50 mm
Catalog # 77577-452
Supplier #:  8644S6-50S
Re-Usable Cleaning Coupons
Catalog # 77577-452
Re-Usable Cleaning Coupons
Re-Usable Cleaning Coupons

Stainless steel 316L - satin, 50×50 mm

Price
$214.00
Log in for availability
Each
Product Details & Documents

Cleaning coupons are great for validating a cleaning regime.

  • All popular sizes available
  • Wide variety of materials and finishes

Sampling Systems extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.

Specifications
  • Description:
    Stainless steel 316L - satin
  • W×L:
    50×50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.