Cleaning Coupons
Cleaning coupon, 316 stainless steel (satin finish), 50×50 mm
Catalog # 77577-450
Supplier: SAMPLING SYSTEMS LTD
Supplier #: 8644S6-50B
Jump to:
Product Details & Documents
These cleaning validation coupons are available in all popular materials and various sizes.
Specifications
- Description:Cleaning coupon, 316 stainless steel (satin finish)
- W×L:50×50 mm