Re-Usable Cleaning Coupons
Polypropylene, 100×100 mm
Catalog # 77577-436
Supplier: SAMPLING SYSTEMS LTD
Supplier #: 8644PVC-100
Jump to:
Product Details & Documents
Cleaning coupons are great for validating a cleaning regime.
- All popular sizes available
- Wide variety of materials and finishes
Sampling Systems extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.
Specifications
- Description:Polypropylene
- W×L:100×100 mm