Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sack Clips

Sack Clips

Sack clip, metal detectable, Acetal, 250 mm, 100 mm
Catalog # 77576-626
Supplier #:  8010M-1
Sack Clips
Catalog # 77576-626
Supplier:  
Sack Clips
Sack Clips

Sack clip, metal detectable, Acetal, 250 mm, 100 mm

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SACK CLIP METAL DETACTEBALE Catalog #78117-069

Product 78117-069 is a replacement for 77576-626 but varies by Brand, Packaging or Quantity.

Product Details & Documents

Sack clips are designed for the easy closing and sealing of bags.

  • Enables quick and easy sealing of bags and sacks
  • Heavy-duty design
  • Available in a range of materials
  • Available from stock
  • Fast delivery

The plastic sack clip is perfect for closing and sealing a range of different bags and sacks. Simply insert the bag into the jaws of the clip and click shut.

Specifications
  • Description:
    Sack clip, metal detectable
  • Length:
    250 mm
  • Material:
    Acetal
  • Overall length:
    100 mm