Accupod Solid Samplers
Accupod sampler, non sterile, HDPE, 5 ml
Catalog # 77577-324
Supplier #: 8360T-50
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ACCUPOD 5.0ML HDPE NATURAL Catalog #78117-130
Product 78117-130 is a replacement for 77577-324 but varies by Brand, Packaging or Quantity.
Product Details & Documents
The AccuPod solid sampler is ideal for unit dose sampling of powders and granules.
- Suitable for sampling blend uniformity
- Next generation in sampling technology
- Designed to minimize sampling error
The AccuPod is perfect for use in drug development and is frequently used in sampling blend uniformity. Ideal for sampling free flowing powders and granules. Sample remains in the pod until needed.
Specifications
- Volume:5 ml
- Description:Accupod sampler, non sterile
- Material:HDPE