Dipping Bottles
Stainless steel 304, 500 ml, 207 mm, 62 mm
77576-570
: 5332S-500
Dipping bottles are excellent for sampling in open water, as well as in tanks and drums.
- Wide range available
- Stainless steel 304 or brass construction
- Customisable
Sampling liquids at depth is easy with these dipping bottles. Available in either 304 stainless steel or brass construction, they can take liquid samples from a variety of locations including open water kegs, drums, and wells.
- Ø:62 mm
- Capacity:500 ml
- Length:207 mm
- Material:Stainless steel 304



