Measuring Scoops
5 ml, 193 mm, 22 mm
Catalog # 77576-332
Supplier: SAMPLING SYSTEMS LTD
: 1900A-05
Jump to:
Product Details & Documents
These measuring scoops made from high quality 316 stainless steel are ideal for scooping out fixed volumes of powder.
- Crevice-free
- Polished
Specifications
- Ø:22 mm
- Capacity:5 ml
- Length:193 mm



