ID Labels with Sequential Number and Barcode
Sequential, barcoded ID labels, 250/roll
Catalog # 77577-220
Supplier #: 8226A-01
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ID LABEL SEQUENTIAL NUMBER AND BARCODE Catalog #78117-067
Product 78117-067 is a replacement for 77577-220 but varies by Brand, Packaging or Quantity.
Product Details & Documents
These ID labels are ideal for unique labelling.
- Each label has a pre-printed reference number
- Easily shows when items have been sealed
- Barcode also allows for easy scanning
This range of ID labels features a pre-printed reference number which makes unique labelling simple and easy. They can be attached to boxes, bottles, or tanks, and clearly shows when an item has been sealed. The sequential number is also shown in barcode form so it can be quickly scanned.
Specifications
- Description:Sequential, barcoded ID labels
- Packaging:250/roll