Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Sleeve Sampler

Heavy duty handle, Stainless steel 316L, 1500 mm
  77576-016
 :  1035A-1500
Sleeve Sampler
  77576-016
Sleeve Sampler
Sleeve Sampler

Heavy duty handle, Stainless steel 316L, 1500 mm

 
$336.00
   
Each
 

The sleeve sampler is specially designed for large volumes at great depths.

  • Adjustable length and bright finish design
  • Extremely strong and robust extension rods

Suitable for free-flowing powders and granules.

 
  • Description:
    Heavy duty handle
  • Length:
    1500 mm
  • Material:
    Stainless steel 316L
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.