Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Sleeve Sampler

Heavy duty handle, Stainless steel 316L, 1000 mm
Catalog # 77576-014
 :  1035A-1000
Sleeve Sampler
Catalog # 77576-014
Sleeve Sampler
Sleeve Sampler

Heavy duty handle, Stainless steel 316L, 1000 mm

Price
$302.00
Log in for availability
Each
Product Details & Documents

The sleeve sampler is specially designed for large volumes at great depths.

  • Adjustable length and bright finish design
  • Extremely strong and robust extension rods

Suitable for free-flowing powders and granules.

Specifications
  • Description:
    Heavy duty handle
  • Length:
    1000 mm
  • Material:
    Stainless steel 316L
  • Cat. No.:
    77576-014
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.