Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Aluminium Sampling Case

Aluminium Sampling Case

Aluminium sampling case, 500 mm
Catalog # 77577-394
Supplier #:  8509A-500
Aluminium Sampling Case
Catalog # 77577-394
Supplier:  
Aluminium Sampling Case
Aluminium Sampling Case

Aluminium sampling case, 500 mm

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
ALUMINIUM CASE. 490MM Catalog #78117-024

Product 78117-024 is a replacement for 77577-394 but varies by Brand, Packaging or Quantity.

Product Details & Documents

The aluminium sampling case is designed for storing samplers and equipment.

  • Available in a range of sizes
  • Lightweight and robust design
  • Foam lined to protect equipment when transported
  • Available from stock
  • Fast delivery

The sampling systems range of aluminium cases are perfect for any sampler storage requirements. They are made from robust and lightweight material and include an interior foam lining, ensuring that equipment is kept safe and protected at all times. The cases are lockable and come with a carrying handle.

Specifications
  • Description:
    Aluminium sampling case
  • Interior dimensions:
    500 mm