Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
SteriWare® Disposable Forceps

SteriWare® Disposable Forceps

White, 115 mm
Catalog # 77576-710
Supplier #:  8025H-115
SteriWare® Disposable Forceps
Catalog # 77576-710
Supplier:  
SteriWare® Disposable Forceps
SteriWare® Disposable Forceps

White, 115 mm

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
DISP TWEEZER HDPE Catalog #78117-072

Product 78117-072 is a replacement for 77576-710 but varies by Brand, Packaging or Quantity.

Product Details & Documents

These diposable tweezers made from 100% virgin HDPE are ideal for picking up items up to 25 mm. Its special dual action design means it can pick up both small and large items.

  • Made from FDA and EC conforming material
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • Gamma irradiated (SAL 10⁻⁶)
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)

Packaging: Manufactured and packed in a class 100000 cleanroom environment. Individually bagged in a heat sealed PE bag.

Specifications
  • Color:
    White
  • Length:
    115 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.