CLONE CCR8 NM005201 HU TAG ORF 200UL
Catalog # MSPP-RC5427L2V
Supplier: Origene Technologies
CLONE CCR8 NM005201 HU TAG ORF 200UL
Catalog # MSPP-RC5427L2V
Supplier: Origene Technologies
Supplier Number: RC215427L2V
Frequently Bought Together
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
Pre-Cut Western Blotting Membranes, Azure Biosystems
Supplier: Azure Biosystems
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: PROTEIN SIMPLE MS
Chemi Blot Blocking Buffer, Azure
Supplier: Azure Biosystems
Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Yeast Mating Factor alpha (from Yeast)
Supplier: Genscript
Blue Prestained Protein Standard, Broad Range (11 to 250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
PI MARKER 9.50 200UL
Supplier: PROTEIN SIMPLE MS
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: AB SCIEX LLC MS
Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
DNA BINDING BUFFER 100 ML
Supplier: Zymo Research
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
TrueBlot® IP/Western Blot, Rockland Immunochemicals
Supplier: Rockland Immunochemical