Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Cryogenic Printer Labels, Brady

Printer labels, Polyester, White, BBP®33, 6,4×44,5 mm (0.25×1.75"), 1 label/row
Catalog # 77574-932
Supplier:   Brady Worldwide
 :  133961
Cryogenic Printer Labels, Brady
Catalog # 77574-932
Supplier:  Brady Worldwide
Cryogenic Printer Labels, Brady
Cryogenic Printer Labels, Brady

Printer labels, Polyester, White, BBP®33, 6,4×44,5 mm (0.25×1.75"), 1 label/row

Price
$108.98
Log in for availability
Roll of 1
Product Details & Documents

Cryogenic labels for tubes, vials, and canes. Chemical- and stain-resistant labels for slides and cassettes. General labels for straws, petri dishes, bottles, flasks, microplates and bags.

Cryogenic labels: temperatures from –196 to 121 °C (–321 to 250 °F) and hot water baths.
B-461 Ultra-thin label with clear tail for tubes.
B-490 Freezerbondz® label for frozen tubes.
B-492 Freezerbondz® ultra-thin for frosted/frozen tubes and canes.

General labels:
B-424 Economical paper label for general purpose.
B-427 Straw label for wrapping; working temperatures from –196 to 80 °C (–321 to 176 °F).
B-430 Petri dish and bag label; working temperatures from –80 to 121 °C (–112 to 250 °F).
B-432 Removable label for all room temperature applications.

Specifications
  • For Use With:
    BBP®33
  • Color:
    White
  • Description:
    Printer labels
  • Dimensions:
    6,4×44,5 mm (0.25×1.75")
  • Material:
    Polyester
  • Packaging:
    1 label/row