Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Cleanroom Sticky Notes

Cleanroom Sticky Notes

Class 10 cleanroom sticky notes, 7.6×7.6 cm (3×3"), Green
Catalog # 77574-778
Supplier #:  POST-IT-GRE
Cleanroom Sticky Notes
Catalog # 77574-778
Cleanroom Sticky Notes
Cleanroom Sticky Notes

Class 10 cleanroom sticky notes, 7.6×7.6 cm (3×3"), Green

Price
$173.37
Log in for availability
Case of 24
Product Details & Documents

Class 10 cleanroom sticky notes include 100 latex sheets per pad and are designed for use in critical environments where low particulation and ESD's are required.

  • 28# latex paper
  • 3×3" note pads
  • 24 pads per case

The latex sheets are smudge-resistant and the adhesive used for binding the sheets does not emit ESD's or carry particulates when a sheet is removed.

Documents

Specifications
  • Color:
    Green
  • Description:
    Class 10 cleanroom sticky notes
  • W×L:
    7.6×7.6 cm (3×3")