$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
BUFFER NEUTRALIZE AND GLOVE 10ML PK100
MSPP-MMMPSSL10
:
NEOGEN CORP (MI) MS
BUFFER NEUTRALIZE AND GLOVE 10ML PK100
MSPP-MMMPSSL10
:
NEOGEN CORP (MI) MS
:
MMMPSSL10NB