Orders LinkContactUsLinkComponent
Triangles
Triangles
Triangles, iron wire, 5.1 cm (2")
Catalog # 21750-095
Supplier #:  H-23860
Triangles
Catalog # 21750-095
Triangles
Triangles
Triangles, iron wire, 5.1 cm (2")
Price
$3.19
Log in for availability
Each
Product Details & Documents
These triangles are made up of iron wire (16 gauge) galvanized with plain clay pipe stems.
Smooth pipe stem triangles protect glassware on hotplates and over burner flames.
Specifications
  • Description:
    Triangles, iron wire, 5.1 cm (2")