Objectives for REVOLVE Microscopes, Echo
Plan fluorite, 17 mm, 0.13 NA
Catalog # 77529-394
Supplier: DISCOVER ECHO INC.
: CMAG0042
Jump to:
Product Details & Documents
Specifications
- Magnification:4×
- Description:Plan fluorite
- Aperture:0.13 NA
- Working distance:17 mm