Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TAG PLASTIC 4-3/4X2-3/8IN RED PK100
Catalog # MSPP-S-5544RPW
Supplier:
ULINE MS
Expand image
TAG PLASTIC 4-3/4X2-3/8IN RED PK100
Catalog # MSPP-S-5544RPW
Supplier:
ULINE MS
Supplier Number:
S-5544R-PW