Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
APS/TEMED

APS/TEMED

25 ml
Catalog # 77510-446
Flammable
Corrosive
Toxic
Supplier:   RPI
Supplier #:  T18000-0.025
APS/TEMED
Catalog # 77510-446
Supplier:  RPI
CAS Number:  7727-54-0/110-18-9
APS/TEMED
APS/TEMED

25 ml

CAS Number:  7727-54-0/110-18-9
Price
$34.16
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Tertiary amine used to catalyze formation of free radicals from ammonium persulfate and riboflavin. Suitable for electrophoresis and tested as a catalyst for polyacrylamide gel electrophoresis.

Caution: Research or further manufacturing use only, not for food or drug use.

Synonyms:  Ammonium persulphate/N,N,N′,N′-Tetramethylethylenediamine, Ammonium peroxodisulphate/N,N,N′,N′-Tetramethylethylenediamine
  • CAS Number:7727-54-0/110-18-9
  • Boiling Pt:120...122 °C
  • Melting Pt:–55 °C
  • Flash Pt:20 °C
  • Storage Temperature:Room Temperature
  • EINECS:203-744-6
  • ADR:3,II
Specifications

Specification Test Results

  • Appearance:
    Clear, Colorless Solution
  • Identification:
    Conforms
  • Purity (GC):
    ≥ 97 %
  • BSE/TSE Free:
    Conform

Specifications

  • Size:
    25 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.