Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dextran 70

Dextran 70

100 g
Catalog # 77510-240
Supplier:   RPI
Supplier #:  D30850-100.0
Dextran 70
Catalog # 77510-240
Supplier:  RPI
CAS Number:  9004-54-0
Dextran 70
Dextran 70

100 g

CAS Number:  9004-54-0
Price
$156.76
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caution: Research or further manufacturing use only, not for food or drug use.

  • CAS Number:9004-54-0
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00130935
  • Storage Temperature:Refrigerator
  • EINECS:232-677-5
Specifications

Specification Test Results

  • Appearance:
    White to slightly off white powder
  • Loss on Drying:
    ≤ 7.0 %
  • Ash Content:
    ≤ 2.0 %
  • Specific Rotation[α ]20D (2% solution):
    +195 - +203 °
  • Intrinsic Viscosity @ 37 oC (1% Solution):
    0.22 - 0.27
  • pH (6% Solution):
    4.5 - 7.0
  • Color of solution @ 375nm (6% solution):
    ≤ 0.15 OD
  • Cold water solubility (1% solution):
    Soluble in cold water
  • Non Animal Origin:
    Conforms
  • BSE/TSE Free:
    BSE/TSE Free
  • Country of Origin:
    CA

Specifications

  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.