Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
TE Buffer

TE Buffer

Tris-EDTA, 1X, 1 L
Catalog # 77510-464
Supplier:   RPI
Supplier #:  T35150-1000.0
TE Buffer
Catalog # 77510-464
Supplier:  RPI
CAS Number:  
TE Buffer
TE Buffer

Tris-EDTA, 1X, 1 L

CAS Number:  
Price
$77.82
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

TE buffer, at 1X concentration, is a commonly used solution, particularly in molecular biology and biochemistry. TE stands for Tris-EDTA, as it contains Tris (tris(hydroxymethyl)aminomethane) and EDTA (ethylenediaminetetraacetic acid).

  • It is also used in DNA and RNA quantification assays, such as spectrophotometry and fluorometry

TE buffer provides a stable environment for DNA and RNA by protecting them from degradation. Tris acts as a buffering agent, maintaining a stable pH, while EDTA chelates divalent metal ions, which are cofactors for nucleases, thus inhibiting their activity and protecting nucleic acids from enzymatic degradation.

TE buffer is commonly used in various molecular biology techniques, such as DNA and RNA extraction, storage of nucleic acids, DNA and RNA gel electrophoresis, and PCR (polymerase chain reaction) assays.

Caution: Research or further manufacturing use only, not for food or drug use.

Specifications

Specifications

  • Concentration:
    1X
  • Description:
    Tris-EDTA
  • Size:
    1 L
  • Application:
    Biological buffers
  • Cat. No.:
    77510-464
  • Appearance:
    Colorless liquid
  • Storage Temperature:
    RT

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.