Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER AAV AEX AAV6 ELUTION 21205
Catalog # 77509-548
Supplier:
Teknova
Expand image
BUFFER AAV AEX AAV6 ELUTION 21205
Catalog # 77509-548
Supplier:
Teknova
Supplier Number:
21205